Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAAACATCAACTCACACCAGAATA[C/G]GAGTAACTTTTAGGATAATCACAGG
Species: |
Human | |||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176590 MIM: 609247 | |||||||||||||||||||||||||||||||||||
Literature Links: |
LOC646903 PubMed Links | |||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS - Not Available | African American
|
YRI (Yoruba) - Not Available | ||||||
SAS - Not Available | Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR - Not Available | Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR - Not Available | ||||||||
AMR - Not Available |
LOC646903 - uncharacterized LOC646903 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PFN2 - profilin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002628.4 | Intron | NP_002619.1 | ||||
NM_053024.3 | Intron | NP_444252.1 |
RNF13 - ring finger protein 13 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
Validated |