Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605403 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TLR6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TLR6 - toll like receptor 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006068.4 | 3676 | UTR 3 | NP_006059.2 | |||
XM_005262637.4 | 3676 | UTR 3 | XP_005262694.1 | |||
XM_011513612.2 | 3676 | UTR 3 | XP_011511914.1 | |||
XM_011513613.2 | 3676 | UTR 3 | XP_011511915.1 | |||
XM_011513614.2 | 3676 | UTR 3 | XP_011511916.1 |