Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCGACCCACTTCCAGTGCTGATGA[G/T]GGCTTGAAAAGGTGGTGCTTGGTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605534 MIM: 614248 | ||||||||||||||||||||
Literature Links: |
HMG20A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HMG20A - high mobility group 20A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304504.1 | Intron | NP_001291433.1 | ||||
NM_001304505.1 | Intron | NP_001291434.1 | ||||
NM_018200.3 | Intron | NP_060670.1 | ||||
XM_011521158.2 | Intron | XP_011519460.1 |
PEAK1 - pseudopodium enriched atypical kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024776.3 | Intron | NP_079052.2 |