Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCAGGTAGCTAGTTCTTGGGTCAA[G/T]ATCCCTAGAGAAGGACAGTACCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605534 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HMG20A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
HMG20A - high mobility group 20A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304504.1 | Intron | NP_001291433.1 | ||||
NM_001304505.1 | Intron | NP_001291434.1 | ||||
NM_018200.3 | Intron | NP_060670.1 | ||||
XM_011521158.2 | Intron | XP_011519460.1 |
Set Membership: |
HapMap Validated |