Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612183 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR176 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GPR176 - G protein-coupled receptor 176 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271854.1 | 7309 | Intron | NP_001258783.1 | |||
NM_001271855.1 | 7309 | Intron | NP_001258784.1 | |||
NM_007223.2 | 7309 | Intron | NP_009154.1 | |||
XM_011521167.2 | 7309 | UTR 3 | XP_011519469.1 | |||
XM_011521168.2 | 7309 | UTR 3 | XP_011519470.1 | |||
XM_017021873.1 | 7309 | UTR 3 | XP_016877362.1 | |||
XM_017021874.1 | 7309 | UTR 3 | XP_016877363.1 | |||
XM_017021875.1 | 7309 | UTR 3 | XP_016877364.1 | |||
XM_017021876.1 | 7309 | UTR 3 | XP_016877365.1 | |||
XM_017021877.1 | 7309 | UTR 3 | XP_016877366.1 | |||
XM_017021878.1 | 7309 | UTR 3 | XP_016877367.1 |
Set Membership: |
HapMap |