Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTGAAATTTTCCCAATATAATA[A/C]CTTCCCTACATTCCTCAGACACAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603000 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRPL58 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MRPL58 - mitochondrial ribosomal protein L58 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303265.1 | Intron | NP_001290194.1 | ||||
NM_001545.2 | Intron | NP_001536.1 | ||||
XM_017024574.1 | Intron | XP_016880063.1 |
Set Membership: |
HapMap |