Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGGAGACGTCCCTCTGGAAATGA[C/T]GCTGGAGCTGAATGCTGAGCCCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 103270 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FADS6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FADS6 - fatty acid desaturase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178128.5 | Intron | NP_835229.3 | ||||
XM_005257224.4 | Intron | XP_005257281.2 | ||||
XM_017024458.1 | Intron | XP_016879947.1 |
FDXR - ferredoxin reductase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |