Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTGGGGCACACTGTCTGCATGAC[A/G]AAAGAGGTTGCTTCTGATCCCAGGC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 601159 MIM: 601268 | |||||||||||||||||||||||
Literature Links: |
CCR1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CCR1 - C-C motif chemokine receptor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CCR3 - C-C motif chemokine receptor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164680.1 | Intron | NP_001158152.1 | ||||
NM_001837.3 | Intron | NP_001828.1 | ||||
NM_178328.1 | Intron | NP_847898.1 | ||||
NM_178329.2 | Intron | NP_847899.1 | ||||
XM_006712960.3 | Intron | XP_006713023.1 | ||||
XM_011533335.2 | Intron | XP_011531637.1 | ||||
XM_017005685.1 | Intron | XP_016861174.1 | ||||
XM_017005686.1 | Intron | XP_016861175.1 |