Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTATTTTGACTTTCATTTTATTAG[G/T]GCTTGAAAGGTAAGACATATTTACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603974 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF92 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF92 - zinc finger protein 92 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287532.1 | Intron | NP_001274461.1 | ||||
NM_001287533.1 | Intron | NP_001274462.1 | ||||
NM_001287534.1 | Intron | NP_001274463.1 | ||||
NM_007139.3 | Intron | NP_009070.2 | ||||
NM_152626.3 | Intron | NP_689839.1 |
Set Membership: |
HapMap |