Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 107271 MIM: 609089 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD59 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CD59 - CD59 molecule | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FBXO3 - F-box protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012175.3 | Intron | NP_036307.2 | ||||
NM_033406.2 | Intron | NP_208385.1 | ||||
XM_011519980.1 | Intron | XP_011518282.1 | ||||
XM_011519981.2 | Intron | XP_011518283.1 | ||||
XM_011519982.1 | Intron | XP_011518284.1 | ||||
XM_017017533.1 | Intron | XP_016873022.1 |