Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGTTTCTAAAAAATGCAGAGCAG[C/G]CTTTATTCTTGTTTTCCCAGGTAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 107271 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C11orf91 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C11orf91 - chromosome 11 open reading frame 91 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CD59 - CD59 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000611.5 | 5690 | UTR 3 | NP_000602.1 | |||
NM_001127223.1 | 5690 | UTR 3 | NP_001120695.1 | |||
NM_001127225.1 | 5690 | UTR 3 | NP_001120697.1 | |||
NM_001127226.1 | 5690 | UTR 3 | NP_001120698.1 | |||
NM_001127227.1 | 5690 | UTR 3 | NP_001120699.1 | |||
NM_203329.2 | 5690 | UTR 3 | NP_976074.1 | |||
NM_203330.2 | 5690 | UTR 3 | NP_976075.1 | |||
NM_203331.2 | 5690 | UTR 3 | NP_976076.1 |