Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAATCAAGCAGTTATTATCCATCA[A/T]CCTAGAAACTATTGGGCCTATAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
60 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602411 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EXTL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EXTL2 - exostosin like glycosyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033025.2 | Intron | NP_001028197.1 | ||||
NM_001261440.1 | Intron | NP_001248369.1 | ||||
NM_001261441.1 | Intron | NP_001248370.1 | ||||
NM_001261442.1 | Intron | NP_001248371.1 | ||||
NM_001439.3 | Intron | NP_001430.1 | ||||
XM_005270621.1 | Intron | XP_005270678.1 | ||||
XM_011540994.2 | Intron | XP_011539296.1 | ||||
XM_011540995.2 | Intron | XP_011539297.1 | ||||
XM_011540996.2 | Intron | XP_011539298.1 | ||||
XM_017000651.1 | Intron | XP_016856140.1 |
Set Membership: |
HapMap |