Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGATGCCCAGGAGGGCATGAGTGTC[A/G]TGTTGGATAACCTGCTCTCCAAGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611203 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJC5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DNAJC5 - DnaJ heat shock protein family (Hsp40) member C5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025219.2 | Intron | NP_079495.1 | ||||
XM_011529048.2 | Intron | XP_011527350.1 | ||||
XM_011529049.1 | Intron | XP_011527351.1 |
MIR941-1 - microRNA 941-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR941-2 - microRNA 941-2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR941-3 - microRNA 941-3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR941-4 - microRNA 941-4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR941-5 - microRNA 941-5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |