Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
42 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605960 MIM: 182891 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EXOSC10 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EXOSC10 - exosome component 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001998.2 | Intron | NP_001001998.1 | ||||
NM_002685.3 | Intron | NP_002676.1 | ||||
XM_005263475.2 | Intron | XP_005263532.2 | ||||
XM_017001494.1 | Intron | XP_016856983.1 | ||||
XM_017001495.1 | Intron | XP_016856984.1 | ||||
XM_017001496.1 | Intron | XP_016856985.1 | ||||
XM_017001497.1 | Intron | XP_016856986.1 |
SRM - spermidine synthase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |