Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
78 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605078 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TARDBP PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
TARDBP - TAR DNA binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007375.3 | Intron | NP_031401.1 | ||||
XM_017000863.1 | Intron | XP_016856352.1 | ||||
XM_017000864.1 | Intron | XP_016856353.1 | ||||
XM_017000865.1 | Intron | XP_016856354.1 | ||||
XM_017000866.1 | Intron | XP_016856355.1 | ||||
XM_017000867.1 | Intron | XP_016856356.1 | ||||
XM_017000868.1 | Intron | XP_016856357.1 |