Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611448 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BMS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
BMS1 - BMS1, ribosome biogenesis factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014753.3 | Intron | NP_055568.3 | ||||
XM_005271846.3 | Intron | XP_005271903.1 | ||||
XM_005271848.3 | Intron | XP_005271905.1 | ||||
XM_005271849.3 | Intron | XP_005271906.1 | ||||
XM_011540402.2 | Intron | XP_011538704.1 | ||||
XM_011540403.2 | Intron | XP_011538705.1 |