Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGCGTTACTCTGGAATTCTAGGC[C/T]CACATGGTGTGGGCCTCCCAGCATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606284 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SORCS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SORCS2 - sortilin related VPS10 domain containing receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020777.2 | Intron | NP_065828.2 | ||||
XM_005247987.4 | Intron | XP_005248044.2 | ||||
XM_011513514.2 | Intron | XP_011511816.1 | ||||
XM_011513515.2 | Intron | XP_011511817.1 | ||||
XM_011513516.2 | Intron | XP_011511818.1 | ||||
XM_017008481.1 | Intron | XP_016863970.1 |