Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTATTTTATTTATATGAATGGAG[A/T]CCGGGAATGAAGTAGAAGAGACTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606284 | ||||||||||||||||||||
Literature Links: |
SORCS2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SORCS2 - sortilin related VPS10 domain containing receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020777.2 | Intron | NP_065828.2 | ||||
XM_005247987.4 | Intron | XP_005248044.2 | ||||
XM_011513514.2 | Intron | XP_011511816.1 | ||||
XM_011513515.2 | Intron | XP_011511817.1 | ||||
XM_011513516.2 | Intron | XP_011511818.1 | ||||
XM_017008481.1 | Intron | XP_016863970.1 |