Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAACAGAGTAGGCACAGTGGACCC[C/T]TCACGATCTCAAGAAATGAAGCTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613137 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TSPAN9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TSPAN9 - tetraspanin 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168320.1 | Intron | NP_001161792.1 | ||||
NM_006675.4 | Intron | NP_006666.1 | ||||
XM_011520912.2 | Intron | XP_011519214.1 | ||||
XM_011520913.2 | Intron | XP_011519215.1 |