Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609426 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MINK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
MINK1 - misshapen like kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024937.3 | Intron | NP_001020108.1 | ||||
NM_001321236.1 | Intron | NP_001308165.1 | ||||
NM_015716.4 | Intron | NP_056531.1 | ||||
NM_153827.4 | Intron | NP_722549.2 | ||||
NM_170663.4 | Intron | NP_733763.1 | ||||
XM_006721531.2 | Intron | XP_006721594.1 | ||||
XM_006721532.2 | Intron | XP_006721595.1 | ||||
XM_006721536.2 | Intron | XP_006721599.1 | ||||
XM_011523906.1 | Intron | XP_011522208.1 | ||||
XM_017024704.1 | Intron | XP_016880193.1 | ||||
XM_017024705.1 | Intron | XP_016880194.1 | ||||
XM_017024706.1 | Intron | XP_016880195.1 | ||||
XM_017024707.1 | Intron | XP_016880196.1 | ||||
XM_017024708.1 | Intron | XP_016880197.1 |
Set Membership: |
HapMap Validated |