Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTCACACAGGAGACTTCCATTCA[C/T]ATGGAATCAGATACACAGCGTGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 153337 MIM: 168440 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LAG3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LAG3 - lymphocyte activating 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PTMS - parathymosin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011520987.1 | Intron | XP_011519289.1 |