Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCTGCACGCCGGGACGCCACCTC[C/T]GCCAGCCGCCTCCACCCGCCCCACA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612184 MIM: 606692 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CASKIN1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CASKIN1 - CASK interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020764.3 | 5511 | UTR 3 | NP_065815.1 |
TRAF7 - TNF receptor associated factor 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032271.2 | 5511 | UTR 3 | NP_115647.2 | |||
XM_005255627.4 | 5511 | UTR 3 | XP_005255684.1 | |||
XM_011522700.1 | 5511 | UTR 3 | XP_011521002.1 |
Set Membership: |
HapMap |