Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCCATAGGAATGCCCACCGCCCT[A/G]GCCAGCGTGAGCCTGTGAGCCTGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164762 MIM: 606538 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CRK - CRK proto-oncogene, adaptor protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MYO1C - myosin IC | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080779.1 | Intron | NP_001074248.1 | ||||
NM_001080950.1 | Intron | NP_001074419.1 | ||||
NM_033375.4 | Intron | NP_203693.3 |