Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600387 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BST1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
BST1 - bone marrow stromal cell antigen 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004334.2 | Intron | NP_004325.2 | ||||
XM_005248185.1 | Intron | XP_005248242.1 | ||||
XM_005248186.1 | Intron | XP_005248243.1 | ||||
XM_011513878.2 | Intron | XP_011512180.1 | ||||
XM_011513879.1 | Intron | XP_011512181.1 | ||||
XM_011513881.2 | Intron | XP_011512183.1 | ||||
XM_017008565.1 | Intron | XP_016864054.1 | ||||
XM_017008566.1 | Intron | XP_016864055.1 | ||||
XM_017008567.1 | Intron | XP_016864056.1 |
Set Membership: |
HapMap Validated |