Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATGTAAGAGAGACAAAGTGTTGC[A/G]GGGTTCAGAGAAGGGAGATGACAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600800 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NAB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NAB1 - NGFI-A binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321312.1 | Intron | NP_001308241.1 | ||||
NM_001321313.1 | Intron | NP_001308242.1 | ||||
NM_001321314.1 | Intron | NP_001308243.1 | ||||
NM_001321315.1 | Intron | NP_001308244.1 | ||||
NM_005966.3 | Intron | NP_005957.2 | ||||
XM_005246582.1 | Intron | XP_005246639.1 | ||||
XM_005246583.1 | Intron | XP_005246640.1 | ||||
XM_005246586.1 | Intron | XP_005246643.1 | ||||
XM_005246587.1 | Intron | XP_005246644.1 | ||||
XM_011511219.2 | Intron | XP_011509521.1 | ||||
XM_017004170.1 | Intron | XP_016859659.1 | ||||
XM_017004171.1 | Intron | XP_016859660.1 | ||||
XM_017004172.1 | Intron | XP_016859661.1 | ||||
XM_017004173.1 | Intron | XP_016859662.1 | ||||
XM_017004174.1 | Intron | XP_016859663.1 | ||||
XM_017004175.1 | Intron | XP_016859664.1 | ||||
XM_017004176.1 | Intron | XP_016859665.1 |