Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTACCAATACAGGCCTGCTTGTAT[C/T]GCTTTCCAGCTTTGAAACTTCCATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607333 MIM: 607334 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
STRADB PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
STRADB - STE20-related kinase adaptor beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206864.1 | Intron | NP_001193793.1 | ||||
NM_018571.5 | Intron | NP_061041.2 | ||||
XM_005246669.1 | Intron | XP_005246726.1 | ||||
XM_017004438.1 | Intron | XP_016859927.1 |
TRAK2 - trafficking kinesin protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |