Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600280 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUBP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NUBP1 - nucleotide binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278506.1 | Intron | NP_001265435.1 | ||||
NM_001323594.1 | Intron | NP_001310523.1 | ||||
NM_001323595.1 | Intron | NP_001310524.1 | ||||
NM_001323596.1 | Intron | NP_001310525.1 | ||||
NM_001323597.1 | Intron | NP_001310526.1 | ||||
NM_002484.3 | Intron | NP_002475.2 | ||||
XM_017023252.1 | Intron | XP_016878741.1 |
TVP23A - trans-golgi network vesicle protein 23 homolog A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079512.3 | Intron | NP_001072980.1 | ||||
NM_001318873.1 | Intron | NP_001305802.1 | ||||
XM_006720944.3 | Intron | XP_006721007.1 | ||||
XM_011522654.2 | Intron | XP_011520956.1 | ||||
XM_017023649.1 | Intron | XP_016879138.1 | ||||
XM_017023650.1 | Intron | XP_016879139.1 | ||||
XM_017023651.1 | Intron | XP_016879140.1 |