Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATACTGCTTCAATATCACATACTTG[A/C]AGCTTACACATTCAAGAATAATTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
VWDE PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
VWDE - von Willebrand factor D and EGF domains | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135924.1 | Intron | NP_001129396.1 | ||||
XM_005249641.1 | Intron | XP_005249698.1 | ||||
XM_011515174.1 | Intron | XP_011513476.1 | ||||
XM_011515175.1 | Intron | XP_011513477.1 | ||||
XM_017011828.1 | Intron | XP_016867317.1 | ||||
XM_017011829.1 | Intron | XP_016867318.1 | ||||
XM_017011830.1 | Intron | XP_016867319.1 |