Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616015 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF180 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RNF180 - ring finger protein 180 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001113561.2 | Intron | NP_001107033.1 | ||||
NM_001323291.1 | Intron | NP_001310220.1 | ||||
NM_001323292.1 | Intron | NP_001310221.1 | ||||
NM_178532.4 | Intron | NP_848627.1 | ||||
XM_017009383.1 | Intron | XP_016864872.1 | ||||
XM_017009384.1 | Intron | XP_016864873.1 | ||||
XM_017009385.1 | Intron | XP_016864874.1 | ||||
XM_017009386.1 | Intron | XP_016864875.1 | ||||
XM_017009387.1 | Intron | XP_016864876.1 | ||||
XM_017009388.1 | Intron | XP_016864877.1 | ||||
XM_017009389.1 | Intron | XP_016864878.1 |
Set Membership: |
HapMap |