Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGCATTCGATAGTATGTAAATGC[A/C]CATAAATAAGAATATTGAGAAGAAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602163 | ||||||||||||||||||||
Literature Links: |
UBE2E2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
UBE2E2 - ubiquitin conjugating enzyme E2 E2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152653.3 | Intron | NP_689866.1 | ||||
XM_005265432.2 | Intron | XP_005265489.1 | ||||
XM_005265433.4 | Intron | XP_005265490.1 | ||||
XM_011534074.2 | Intron | XP_011532376.1 | ||||
XM_011534075.1 | Intron | XP_011532377.1 | ||||
XM_011534076.2 | Intron | XP_011532378.1 | ||||
XM_017007126.1 | Intron | XP_016862615.1 |
UBE2E2-AS1 - UBE2E2 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |