Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTTTTCTATAGCTTCCACTGAGA[A/G]GATCTAATTAGCATTTTAGTCTCCC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 609938 | |||||||||||||||||||||||
Literature Links: |
CADM2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CADM2 - cell adhesion molecule 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167674.1 | Intron | NP_001161146.1 | ||||
NM_001167675.1 | Intron | NP_001161147.1 | ||||
NM_001256502.1 | Intron | NP_001243431.1 | ||||
NM_001256503.1 | Intron | NP_001243432.1 | ||||
NM_001256504.1 | Intron | NP_001243433.1 | ||||
NM_001256505.1 | Intron | NP_001243434.1 | ||||
NM_153184.3 | Intron | NP_694854.2 | ||||
XM_006713081.3 | Intron | XP_006713144.1 | ||||
XM_006713082.3 | Intron | XP_006713145.1 | ||||
XM_017006062.1 | Intron | XP_016861551.1 | ||||
XM_017006063.1 | Intron | XP_016861552.1 |