Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTGACTGAAATAGAGAGCAAACA[A/G]CTCGATGATTAAGAGAAGTTTTGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608288 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IGF2BP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IGF2BP1 - insulin like growth factor 2 mRNA binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160423.1 | Intron | NP_001153895.1 | ||||
NM_006546.3 | Intron | NP_006537.3 | ||||
XM_005256955.3 | Intron | XP_005257012.2 | ||||
XM_011524201.2 | Intron | XP_011522503.1 | ||||
XM_017024022.1 | Intron | XP_016879511.1 |