Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAAGAGCTGAGACATGAGAAAAGA[C/G]GCACTTACACACTAGCACTGGAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604986 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BRAP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BRAP - BRCA1 associated protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006768.4 | Intron | NP_006759.3 | ||||
XM_005253944.4 | Intron | XP_005254001.1 | ||||
XM_011538789.2 | Intron | XP_011537091.1 | ||||
XM_017019992.1 | Intron | XP_016875481.1 | ||||
XM_017019993.1 | Intron | XP_016875482.1 | ||||
XM_017019994.1 | Intron | XP_016875483.1 | ||||
XM_017019995.1 | Intron | XP_016875484.1 |