Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603703 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RPL6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
RPL6 - ribosomal protein L6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000970.4 | Intron | NP_000961.2 | ||||
NM_001024662.2 | Intron | NP_001019833.1 | ||||
NM_001320137.1 | Intron | NP_001307066.1 | ||||
NM_001320138.1 | Intron | NP_001307067.1 | ||||
NM_001320139.1 | Intron | NP_001307068.1 | ||||
NM_001320140.1 | Intron | NP_001307069.1 | ||||
NM_001320141.1 | Intron | NP_001307070.1 | ||||
NM_001320142.1 | Intron | NP_001307071.1 | ||||
XM_017019781.1 | Intron | XP_016875270.1 | ||||
XM_017019782.1 | Intron | XP_016875271.1 |
Set Membership: |
HapMap Validated |