Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600294 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADCY6 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
CHB (Han Chinese)
|
|||||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
ADCY6 - adenylate cyclase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015270.4 | Intron | NP_056085.1 | ||||
XM_006719210.3 | Intron | XP_006719273.1 | ||||
XM_017018743.1 | Intron | XP_016874232.1 |
LINC00935 - long intergenic non-protein coding RNA 935 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4701 - microRNA 4701 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |