Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGGGATAGTGAGAAGACACAGGA[G/T]AAAAAGCCAGTTAGAGAGCTCATCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607929 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CCM2 - CCM2 scaffolding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001029835.2 | Intron | NP_001025006.1 | ||||
NM_001167934.1 | Intron | NP_001161406.1 | ||||
NM_001167935.1 | Intron | NP_001161407.1 | ||||
NM_031443.3 | Intron | NP_113631.1 | ||||
XM_006715785.3 | Intron | XP_006715848.1 | ||||
XM_006715786.3 | Intron | XP_006715849.1 | ||||
XM_011515561.2 | Intron | XP_011513863.1 | ||||
XM_011515562.1 | Intron | XP_011513864.1 | ||||
XM_011515563.2 | Intron | XP_011513865.1 | ||||
XM_011515564.2 | Intron | XP_011513866.1 | ||||
XM_017012671.1 | Intron | XP_016868160.1 | ||||
XM_017012672.1 | Intron | XP_016868161.1 | ||||
XM_017012673.1 | Intron | XP_016868162.1 |