Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603917 MIM: 614905 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EIF3B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
EIF3B - eukaryotic translation initiation factor 3 subunit B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037283.1 | Intron | NP_001032360.1 | ||||
NM_003751.3 | Intron | NP_003742.2 | ||||
XM_011515599.1 | Intron | XP_011513901.1 | ||||
XM_011515600.1 | Intron | XP_011513902.1 | ||||
XM_011515601.1 | Intron | XP_011513903.1 | ||||
XM_017012752.1 | Intron | XP_016868241.1 | ||||
XM_017012753.1 | Intron | XP_016868242.1 |
SNX8 - sorting nexin 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |