Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603917 MIM: 614905 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EIF3B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
EIF3B - eukaryotic translation initiation factor 3 subunit B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037283.1 | 122 | Intron | NP_001032360.1 | |||
NM_003751.3 | 122 | Intron | NP_003742.2 | |||
XM_011515599.1 | 122 | UTR 5 | XP_011513901.1 | |||
XM_011515600.1 | 122 | UTR 5 | XP_011513902.1 | |||
XM_011515601.1 | 122 | UTR 5 | XP_011513903.1 | |||
XM_017012752.1 | 122 | UTR 5 | XP_016868241.1 | |||
XM_017012753.1 | 122 | UTR 5 | XP_016868242.1 |
SNX8 - sorting nexin 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013321.3 | 122 | Intron | NP_037453.1 | |||
XM_011515329.2 | 122 | Intron | XP_011513631.1 | |||
XM_011515330.1 | 122 | Intron | XP_011513632.1 | |||
XM_017012083.1 | 122 | Intron | XP_016867572.1 | |||
XM_017012084.1 | 122 | Intron | XP_016867573.1 |
Set Membership: |
Validated |