Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCATGTTGGAGGAAGGGAGATGT[C/T]GTCTCAGGTGTGTATGAGTGAGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607611 MIM: 608076 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLSCR3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PLSCR3 - phospholipid scramblase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM256-PLSCR3 - TMEM256-PLSCR3 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNK1 - tyrosine kinase non receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001251902.1 | Intron | NP_001238831.1 | ||||
NM_003985.4 | Intron | NP_003976.2 | ||||
XM_011524045.2 | Intron | XP_011522347.1 | ||||
XM_011524046.2 | Intron | XP_011522348.1 | ||||
XM_011524047.2 | Intron | XP_011522349.1 | ||||
XM_011524048.1 | Intron | XP_011522350.1 |