Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTGAAGCTCGCAAGTGCCCACCC[C/T]GCTCGCTGGTGAAGCTGTGTCCGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 164765 | ||||||||||||||||||||
Literature Links: |
CTTN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CTTN - cortactin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184740.1 | Intron | NP_001171669.1 | ||||
NM_005231.3 | Intron | NP_005222.2 | ||||
NM_138565.2 | Intron | NP_612632.1 | ||||
XM_006718447.2 | Intron | XP_006718510.1 | ||||
XM_006718448.3 | Intron | XP_006718511.1 | ||||
XM_017017312.1 | Intron | XP_016872801.1 |