Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614239 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM109A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FAM109A - family with sequence similarity 109 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177996.1 | 3070 | UTR 3 | NP_001171467.1 | |||
NM_001177997.1 | 3070 | UTR 3 | NP_001171468.1 | |||
NM_144671.4 | 3070 | UTR 3 | NP_653272.2 | |||
XM_006719257.3 | 3070 | Intron | XP_006719320.1 | |||
XM_011537976.2 | 3070 | Intron | XP_011536278.1 | |||
XM_017018883.1 | 3070 | Intron | XP_016874372.1 |
Set Membership: |
HapMap |