Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602520 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP2K5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
MAP2K5 - mitogen-activated protein kinase kinase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206804.1 | Intron | NP_001193733.1 | ||||
NM_002757.3 | Intron | NP_002748.1 | ||||
NM_145160.2 | Intron | NP_660143.1 | ||||
XM_011521787.2 | Intron | XP_011520089.1 | ||||
XM_011521788.2 | Intron | XP_011520090.1 | ||||
XM_017022414.1 | Intron | XP_016877903.1 |
Set Membership: |
HapMap Validated |