Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGAATGTTCTTTGGGTTTTTCCA[G/T]TGGATCCTTCACAAGCTCCATTTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607870 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
UNC5B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
UNC5B - unc-5 netrin receptor B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244889.1 | Intron | NP_001231818.1 | ||||
NM_170744.4 | Intron | NP_734465.2 | ||||
XM_011539453.1 | Intron | XP_011537755.1 | ||||
XM_017015834.1 | Intron | XP_016871323.1 | ||||
XM_017015835.1 | Intron | XP_016871324.1 |
UNC5B-AS1 - UNC5B antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |