Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607176 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CALN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CALN1 - calneuron 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017440.2 | 8315 | UTR 3 | NP_001017440.1 | |||
NM_031468.3 | 8315 | UTR 3 | NP_113656.2 | |||
XM_011516594.2 | 8315 | UTR 3 | XP_011514896.1 | |||
XM_011516595.1 | 8315 | UTR 3 | XP_011514897.1 | |||
XM_011516596.2 | 8315 | UTR 3 | XP_011514898.1 | |||
XM_011516597.2 | 8315 | UTR 3 | XP_011514899.1 | |||
XM_011516599.1 | 8315 | UTR 3 | XP_011514901.1 | |||
XM_017012675.1 | 8315 | UTR 3 | XP_016868164.1 | |||
XM_017012676.1 | 8315 | UTR 3 | XP_016868165.1 | |||
XM_017012677.1 | 8315 | UTR 3 | XP_016868166.1 | |||
XM_017012678.1 | 8315 | UTR 3 | XP_016868167.1 | |||
XM_017012679.1 | 8315 | UTR 3 | XP_016868168.1 | |||
XM_017012680.1 | 8315 | UTR 3 | XP_016868169.1 | |||
XM_017012681.1 | 8315 | UTR 3 | XP_016868170.1 | |||
XM_017012682.1 | 8315 | UTR 3 | XP_016868171.1 | |||
XM_017012683.1 | 8315 | UTR 3 | XP_016868172.1 |