Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SNX31 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SNX31 - sorting nexin 31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152628.3 | Intron | NP_689841.3 | ||||
XM_005250815.3 | Intron | XP_005250872.1 | ||||
XM_011516899.1 | Intron | XP_011515201.1 | ||||
XM_017013153.1 | Intron | XP_016868642.1 | ||||
XM_017013154.1 | Intron | XP_016868643.1 | ||||
XM_017013155.1 | Intron | XP_016868644.1 | ||||
XM_017013156.1 | Intron | XP_016868645.1 | ||||
XM_017013157.1 | Intron | XP_016868646.1 | ||||
XM_017013158.1 | Intron | XP_016868647.1 | ||||
XM_017013159.1 | Intron | XP_016868648.1 | ||||
XM_017013160.1 | Intron | XP_016868649.1 | ||||
XM_017013161.1 | Intron | XP_016868650.1 | ||||
XM_017013162.1 | Intron | XP_016868651.1 | ||||
XM_017013163.1 | Intron | XP_016868652.1 | ||||
XM_017013164.1 | Intron | XP_016868653.1 | ||||
XM_017013165.1 | Intron | XP_016868654.1 |
Set Membership: |
HapMap |