Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCCAGCATCAGGCTCTGTCTTGC[A/G]TAGTCAGGCCTAGTGGGGAAGTGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611211 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RELT PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RELT - RELT tumor necrosis factor receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032871.3 | Intron | NP_116260.2 | ||||
NM_152222.1 | Intron | NP_689408.1 | ||||
XM_005274356.2 | Intron | XP_005274413.1 | ||||
XM_006718714.2 | Intron | XP_006718777.1 | ||||
XM_011545305.1 | Intron | XP_011543607.1 |
Set Membership: |
Validated |