Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608894 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AHI1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
AHI1 - Abelson helper integration site 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134830.1 | Intron | NP_001128302.1 | ||||
NM_001134831.1 | Intron | NP_001128303.1 | ||||
NM_001134832.1 | Intron | NP_001128304.1 | ||||
NM_017651.4 | Intron | NP_060121.3 | ||||
XM_011535910.2 | Intron | XP_011534212.1 | ||||
XM_011535911.2 | Intron | XP_011534213.1 | ||||
XM_017010977.1 | Intron | XP_016866466.1 | ||||
XM_017010978.1 | Intron | XP_016866467.1 | ||||
XM_017010979.1 | Intron | XP_016866468.1 | ||||
XM_017010980.1 | Intron | XP_016866469.1 | ||||
XM_017010981.1 | Intron | XP_016866470.1 | ||||
XM_017010982.1 | Intron | XP_016866471.1 | ||||
XM_017010983.1 | Intron | XP_016866472.1 | ||||
XM_017010984.1 | Intron | XP_016866473.1 |