Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606181 MIM: 611320 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDX24 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DDX24 - DEAD-box helicase 24 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IFI27L1 - interferon alpha inducible protein 27 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145249.2 | Intron | NP_660292.1 | ||||
NM_206949.2 | Intron | NP_996832.1 | ||||
XM_005267314.4 | Intron | XP_005267371.1 | ||||
XM_011536405.2 | Intron | XP_011534707.1 | ||||
XM_011536406.1 | Intron | XP_011534708.1 | ||||
XM_011536407.1 | Intron | XP_011534709.1 | ||||
XM_017020961.1 | Intron | XP_016876450.1 | ||||
XM_017020962.1 | Intron | XP_016876451.1 | ||||
XM_017020963.1 | Intron | XP_016876452.1 |