Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATGTTCAGCAGGGATCCCGGACC[C/T]CTATCCACTGGGTACCAATAGTATT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606885 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACADS PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ACADS - acyl-CoA dehydrogenase, C-2 to C-3 short chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000017.3 | Intron | NP_000008.1 | ||||
NM_001302554.1 | Intron | NP_001289483.1 |
MIR4700 - microRNA 4700 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UNC119B - unc-119 lipid binding chaperone B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |