Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCAGCTTGTTGCAACATTGCGCTT[A/G]CCAGGTTCCTGAGAAAGGCATTTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605563 MIM: 613802 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CABP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CABP1 - calcium binding protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MLEC - malectin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303627.1 | 1239 | Intron | NP_001290556.1 | |||
NM_001303628.1 | 1239 | Intron | NP_001290557.1 | |||
NM_014730.3 | 1239 | Intron | NP_055545.1 | |||
XM_011539031.1 | 1239 | UTR 5 | XP_011537333.1 | |||
XM_011539032.1 | 1239 | UTR 5 | XP_011537334.1 |